The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014540.3 | NP_001014540.2 | 4 - 5 | 2063 | 71 |
NM_001014542.3 | NP_001014542.1 | 4 - 5 | 2063 | 71 | |
NM_001169737.2 | NP_001163208.1 | 4 - 5 | 2160 | 71 | |
NM_001169738.2 | NP_001163209.1 | 4 - 5 | 2160 | 71 | |
NM_001169739.2 | NP_001163210.1 | 4 - 5 | 2063 | 71 | |
NM_001169740.2 | NP_001163211.1 | 4 - 5 | 2204 | 71 | |
NM_206171.4 | NP_995893.1 | 4 - 5 | 1962 | 71 | |
NM_206172.3 | NP_995894.2 | 4 - 5 | 2063 | 71 | |
NM_206177.3 | NP_995899.2 | 4 - 5 | 2063 | 71 | |
NM_206178.3 | NP_995900.1 | 4 - 5 | 2063 | 71 | |
GenBank mRNA | AF258462.1 | - | 4 - 5 | 2038 | 71 |
AF387635.1 | - | 4 - 5 | 2038 | 71 | |
AF387636.1 | - | 3 - 4 | 1980 | 71 | |
BT009940.1 | - | 4 - 5 | 2112 | 71 | |
BT011020.1 | - | 4 - 5 | 2058 | 71 |