Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sif
Entrez Gene ID
43892
Gene Name
still life
Gene Aliases
CG32414, CG34418, CG5256, CG5406, Dm_3L:26493, Dmel\CG34418, Dmel_CG34418, RhoGEF, SIF, Sif, Stilllife, lincRNA.447, slf
Chromosome Location
Chr.3L: 5657913 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104046.4 NP_001097516.3 10 - 11 5426 126
NM_001104047.5 NP_001097517.1 21 - 22 5186 126
NM_001104048.2 NP_001097518.2 23 - 24 4846 126
NM_001104049.4 NP_001097519.2 22 - 23 4641 126
NM_001274517.3 NP_001261446.1 22 - 23 5243 126
NM_001274518.2 NP_001261447.1 9 - 10 2114 126
NM_001274519.3 NP_001261448.1 22 - 23 5216 126
NM_001300018.2 NP_001286947.1 21 - 22 6198 126
NM_001347763.1 NP_001334674.1 23 - 24 5162 126
NM_001347764.1 NP_001334675.1 23 - 24 5198 126
NM_079908.3 NP_524647.2 10 - 11 5426 126
NM_168126.4 NP_729084.1 21 - 22 4196 126
NM_206278.2 NP_996000.1 10 - 11 5426 126
GenBank mRNA AY118305.1 - 10 - 11 5404 126

Gene Symbol
CG46320
Entrez Gene ID
30522409
Gene Name
CG46320-PA
Gene Aliases
Dmel\CG46320, Dmel_CG46320, sif-5'utr
Chromosome Location
Chr.3L: 5664121 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347801.1 NP_001334709.1 21 - 22 5186 126
NM_001347802.1 NP_001334710.1 23 - 24 5162 126

View Full Product Details