Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sif
- Entrez Gene ID
-
43892
- Gene Name
- still life
- Gene Aliases
-
CG32414, CG34418, CG5256, CG5406, Dm_3L:26493, Dmel\CG34418, Dmel_CG34418, RhoGEF, SIF, Sif, Stilllife, lincRNA.447, slf
- Chromosome Location
-
Chr.3L: 5657913 - 5749599 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0085447
- Gene Symbol
- CG46320
- Entrez Gene ID
-
30522409
- Gene Name
- CG46320-PA
- Gene Aliases
-
Dmel\CG46320, Dmel_CG46320, sif-5'utr
- Chromosome Location
-
Chr.3L: 5664121 - 5749599 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0284236
View Full Product Details