Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp47
Entrez Gene ID
38644
Gene Name
Ubiquitin specific protease 47
Gene Aliases
BcDNA:LD12764, CG5486, D-UBP-64E, D-Ubp-64E, Dmel\CG5486, Dmel_CG5486, E(var)1, E(var)3-64E, E-var(3)64E, Q24574, UBP, UBP64E, Ubp64, Ubp64E, Upd64E, dUSP47, dUbp64, ubp64, ubp64E, usp47
Chromosome Location
Chr.3L: 5752973 - 5761886 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016756
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079213.4 NP_523937.2 3 - 4 1974 95
NM_168129.4 NP_729087.1 3 - 4 2147 95
NM_206279.4 NP_996001.2 5 - 6 2165 95
GenBank mRNA BT003784.1 - 3 - 4 2127 95

View Full Product Details