Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clk
Entrez Gene ID
38872
Gene Name
Clock
Gene Aliases
CG7391, CLK, CLOCK, Dmel\CG7391, Dmel_CG7391, Jerk, Jrk, PAS1, bHLHe10, clk, clock, dCLK, dCLK/JRK, dCLOCK, dClck, dClk, dClock, dclk, jrk
Chromosome Location
Chr.3L: 7763233 - 7775603 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023076
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014574.2 NP_001014574.1 7 - 8 1945 74
NM_001014576.2 NP_001014576.1 7 - 8 1947 74
NM_079240.3 NP_523964.2 7 - 8 1935 74
NM_170628.2 NP_729294.2 4 - 5 1278 74
NM_206299.2 NP_996021.2 4 - 5 1278 74

View Full Product Details