Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ldbr
Entrez Gene ID
38900
Gene Name
lariat debranching enzyme
Gene Aliases
CG7942, Dmel\CG7942, Dmel_CG7942, Lbdr, Ldbr
Chromosome Location
Chr.3L: 8121827 - 8123615 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035838
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206300.2 NP_996022.1 1 - 2 290 103
GenBank mRNA AY119584.1 - 1 - 2 275 103

View Full Product Details