The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274391.1 | NP_001261320.1 | 9 - 10 | 3034 | 67 |
NM_138984.4 | NP_620469.2 | 9 - 10 | 3028 | 67 | |
GenBank mRNA | BT053678.1 | - | 9 - 10 | 3026 | 67 |