Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- fbl
- Entrez Gene ID
-
46234
- Gene Name
- fumble
- Gene Aliases
-
CG5725, Dmel\CG5725, Dmel_CG5725, Fbl, PANK, dPANK, dPANK/Fbl, dPanK
- Chromosome Location
-
Chr.3L: 20384429 - 20391913 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011205
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138997.2 |
NP_620550.1 |
1 - 2 |
389 |
71 |
GenBank mRNA |
BT015256.1 |
- |
1 - 2 |
389 |
71 |
View Full Product Details