Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cg
Entrez Gene ID
36571
Gene Name
combgap
Gene Aliases
CG, CG8367, Cg, Dmel\CG8367, Dmel_CG8367, l(2)07659, l(2)k11504, mig
Chromosome Location
Chr.2R: 14173693 - 14182838 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000289
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103834.4 NP_001097304.2 4 - 5 1404 101
NM_001144194.2 NP_001137666.1 4 - 5 1404 101
NM_166034.2 NP_725362.1 4 - 5 1404 101

View Full Product Details