Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATPCL
Entrez Gene ID
36760
Gene Name
ATP citrate lyase
Gene Aliases
ACL, BcDNA:LD21334, CG8322, DmATPCL, Dmel\CG8322, Dmel_CG8322, anon-WO0140519.179, dATPCL, l(2)01466, l(2)k09217, n(2)k09217
Chromosome Location
Chr.2R: 15993531 - 16001290 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299541.1 NP_001286470.1 5 - 6 1597 76

View Full Product Details