Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sd
Entrez Gene ID
32536
Gene Name
scalloped
Gene Aliases
CG8544, DROEXO, DROEXO2, Dmel\CG8544, Dmel_CG8544, EP(X)1088, EP1088, SD, Sd, TEAD, anon-EST:Liang-2.14, clone 2.14, l(1)G0239, l(1)G0262, l(1)G0309, l(1)G0315, l(1)G0483, l(1)HF394, l(1)III, sp
Chromosome Location
Chr.X: 15804370 - 15827682 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003345
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103519.3 NP_001096989.1 3 - 4 1084 94
NM_001103520.3 NP_001096990.1 4 - 5 1033 94
NM_001103521.3 NP_001096991.1 4 - 5 828 94
NM_001103522.3 NP_001096992.1 4 - 5 920 94
NM_001169290.2 NP_001162761.1 4 - 5 1033 94
NM_001169291.2 NP_001162762.1 4 - 5 1076 94
NM_001258767.2 NP_001245696.1 4 - 5 1841 94
NM_001258768.1 NP_001245697.1 4 - 5 894 94
NM_001258769.2 NP_001245698.1 3 - 4 323 94
NM_001258770.3 NP_001245699.1 4 - 5 1168 94
NM_001258771.3 NP_001245700.1 3 - 4 381 94
NM_001258772.2 NP_001245701.1 4 - 5 1577 94
NM_001258773.2 NP_001245702.1 4 - 5 1151 94
NM_001272652.1 NP_001259581.1 3 - 4 701 94
NM_001272653.2 NP_001259582.1 5 - 6 1084 94
NM_001272654.2 NP_001259583.1 4 - 5 1172 94
NM_001298370.1 NP_001285299.1 3 - 4 447 94
NM_001316637.1 NP_001303566.1 4 - 5 1841 94
NM_001382196.1 NP_001368984.1 4 - 5 1168 94
NM_078614.5 NP_511169.1 5 - 6 975 94
NM_167465.3 NP_727900.1 3 - 4 473 94
NM_206727.3 NP_996450.1 3 - 4 637 94
GenBank mRNA BT133072.1 - 4 - 5 1030 94
M83787.1 - 5 - 6 973 94

View Full Product Details