Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh
Entrez Gene ID
32780
Gene Name
Shaker
Gene Aliases
BcDNA:GH03046, CG12348, CG17860, CG7640, Dmel\CG12348, Dmel_CG12348, EKO, Eko, F-c, K[[v]]1, Kv1, SH, Sha, Shw, V-I, minisleep, sh, sha
Chromosome Location
Chr.X: 17924307 - 18063247 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003380
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169317.1 NP_001162788.1 2 - 3 371 101
NM_001201739.2 NP_001188668.1 4 - 5 2663 101
NM_001258797.1 NP_001245726.1 3 - 4 1306 101
NM_001258798.2 NP_001245727.1 6 - 7 2270 101
NM_001272735.3 NP_001259664.1 4 - 5 2663 101
NM_001272736.2 NP_001259665.2 3 - 4 851 101
NM_001272737.1 NP_001259666.2 3 - 4 1626 101
NM_001316640.1 NP_001303569.1 3 - 4 916 101
NM_078669.5 NP_523393.3 3 - 4 916 101
NM_167592.4 NP_728120.1 3 - 4 450 101
NM_167594.4 NP_728122.1 4 - 5 2663 101
NM_167595.4 NP_728123.1 3 - 4 916 101
NM_167596.5 NP_728124.1 3 - 4 450 101
NM_206774.2 NP_996497.1 3 - 4 859 101
NM_206775.5 NP_996498.2 6 - 7 2270 101
GenBank mRNA BT050432.1 - 3 - 4 450 101
X06184.1 - - 369 101
X07132.1 - - 544 101
X07133.1 - - 248 101
X07134.1 - - 859 101

View Full Product Details