Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frq2
Entrez Gene ID
32799
Gene Name
Frequenin 2
Gene Aliases
CG5907, Dmel\CG5907, Dmel_CG5907, Frq, dNCS-1, frq, frq2
Chromosome Location
Chr.X: 18177612 - 18204407 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0083228
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272746.1 NP_001259675.1 3 - 4 736 79
NM_080354.4 NP_525093.1 3 - 4 541 79
NM_206778.3 NP_996501.1 3 - 4 603 79
NM_206779.3 NP_996502.1 4 - 5 466 79
GenBank mRNA BT133347.1 - 3 - 4 539 79

View Full Product Details