Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iris
Entrez Gene ID
33290
Gene Name
CG4715-PA
Gene Aliases
CG4715, CG4715-env, Dm-cF, Dmel\CG4715, Dmel_CG4715
Chromosome Location
Chr.2L: 1132332 - 1134098 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031305
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134726.3 NP_608570.1 1 - 1 1639 110
GenBank mRNA AY069209.1 - 1 - 1 1639 110

View Full Product Details