Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chinmo
Entrez Gene ID
33343
Gene Name
Chronologically inappropriate morphogenesis
Gene Aliases
CG10871, CG17156, CG17649, CG31666, Chinmo, Dmel\CG31666, Dmel_CG31666, F, l(2)04111
Chromosome Location
Chr.2L: 1651258 - 1704682 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086758
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201751.2 NP_001188680.1 4 - 5 2369 64
NM_001258910.2 NP_001245839.1 4 - 5 2369 64
NM_134766.6 NP_608610.1 3 - 4 988 64
NM_164428.3 NP_722717.2 5 - 6 1086 64
NM_164429.3 NP_722718.1 5 - 6 2138 64
NM_164430.4 NP_722719.1 4 - 5 2068 64
GenBank mRNA AY060464.1 - 5 - 6 1095 64
BT050405.1 - 4 - 5 2067 64

View Full Product Details