Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chinmo
Entrez Gene ID
33343
Gene Name
Chronologically inappropriate morphogenesis
Gene Aliases
CG10871, CG17156, CG17649, CG31666, Chinmo, Dmel\CG31666, Dmel_CG31666, F, l(2)04111
Chromosome Location
Chr.2L: 1651258 - 1704682 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086758
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201751.2 NP_001188680.1 6 - 7 3355 57
NM_001258910.2 NP_001245839.1 6 - 7 3355 57
NM_134766.6 NP_608610.1 5 - 6 1974 57
NM_164428.3 NP_722717.2 7 - 8 2072 57
NM_164429.3 NP_722718.1 7 - 8 3124 57
NM_164430.4 NP_722719.1 6 - 7 3054 57
GenBank mRNA AY060464.1 - 7 - 8 2081 57
BT050405.1 - 6 - 7 3053 57

View Full Product Details