Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hydr2
Entrez Gene ID
33532
Gene Name
alpha/beta hydrolase2
Gene Aliases
C24, CG3488, CT11757, DMU29170, Dmel\CG3488, Dmel_CG3488, HYDR2, anon-23D, anon-23Da, c24, cg3488
Chromosome Location
Chr.2L: 3163645 - 3169462 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014906
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258923.2 NP_001245852.1 1 - 2 424 76
NM_134907.4 NP_608751.2 1 - 2 424 76
GenBank mRNA BT010082.1 - 1 - 2 411 76

View Full Product Details