Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
path
Entrez Gene ID
39106
Gene Name
pathetic
Gene Aliases
CG3424, Dmel\CG3424, Dmel_CG3424, Path, Pathetic, anon-WO0118547.271, anon-WO0118547.370, pth
Chromosome Location
Chr.3L: 9494555 - 9505328 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036007
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274705.1 NP_001261634.1 6 - 7 911 83
NM_140070.3 NP_648327.1 5 - 6 561 83
NM_168349.2 NP_729505.2 3 - 4 321 83
GenBank mRNA AY070669.1 - 5 - 6 563 83
AY070688.1 - 5 - 6 563 83
BT088839.1 - 5 - 6 535 83

View Full Product Details