Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
path
Entrez Gene ID
39106
Gene Name
pathetic
Gene Aliases
CG3424, Dmel\CG3424, Dmel_CG3424, Path, Pathetic, anon-WO0118547.271, anon-WO0118547.370, pth
Chromosome Location
Chr.3L: 9494555 - 9505328 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036007
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274705.1 NP_001261634.1 7 - 8 1829 64
NM_140070.3 NP_648327.1 6 - 7 1479 64
NM_168349.2 NP_729505.2 4 - 5 1239 64
GenBank mRNA AY070669.1 - 6 - 7 1481 64
AY070688.1 - 6 - 7 1481 64
BT088839.1 - 6 - 7 1453 64

View Full Product Details