Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
simj
Entrez Gene ID
39212
Gene Name
simjang
Gene Aliases
CG32067, CG7983, CG8000, Dmel\CG32067, Dmel_CG32067, SIMJ, anon-WO0118547.361, dp66, l(3)01814, l(3)j4A5, l(3)rN672, p66, p66/68-like
Chromosome Location
Chr.3L: 10664739 - 10690734 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010762
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274755.1 NP_001261684.1 4 - 5 1923 76
NM_001274756.1 NP_001261685.1 4 - 5 1899 76
NM_140150.3 NP_648407.1 4 - 5 1899 76
NM_168420.2 NP_729624.1 4 - 5 1899 76
NM_168421.2 NP_729625.1 5 - 6 1658 76
GenBank mRNA AY051828.1 - 4 - 5 1864 76

View Full Product Details