Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RhoGAP68F
Entrez Gene ID
39385
Gene Name
Rho GTPase activating protein at 68F
Gene Aliases
CG 6811, CG6811, Dmel\CG6811, Dmel_CG6811, RhoGAP
Chromosome Location
Chr.3L: 12138868 - 12143380 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036257
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274815.1 NP_001261744.1 2 - 3 317 61
NM_001274816.1 NP_001261745.1 2 - 3 251 61
NM_140295.3 NP_648552.1 2 - 3 251 61
GenBank mRNA AY069311.1 - 2 - 3 238 61

View Full Product Details