Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slob
Entrez Gene ID
34038
Gene Name
Slowpoke binding protein
Gene Aliases
CG13792, CG43756, CG6772, Dmel\CG43756, Dmel_CG43756, slob
Chromosome Location
Chr.2L: 7642005 - 7675078 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264087
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038792.1 NP_001033881.1 6 - 7 1574 78
NM_001201781.1 NP_001188710.1 7 - 8 1625 78
NM_001201782.1 NP_001188711.1 8 - 9 1678 78
NM_001201783.1 NP_001188712.1 5 - 6 1406 78
NM_001201784.2 NP_001188713.1 6 - 7 1470 78
NM_001273282.1 NP_001260211.1 6 - 7 1574 78
NM_001273283.1 NP_001260212.1 7 - 8 1833 78
NM_058001.3 NP_477349.1 6 - 7 1574 78
NM_135287.2 NP_609131.2 5 - 6 1249 78
NM_164756.3 NP_723292.1 8 - 9 2001 78
NM_164757.2 NP_723293.1 7 - 8 1638 78
GenBank mRNA BT021228.1 - 6 - 7 1575 78

View Full Product Details