Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
raw
Entrez Gene ID
44851
Gene Name
CG12437-PA
Gene Aliases
CG12437, CG9321, Dmel\CG12437, Dmel_CG12437, Raw, cyr, ensh, l(2)06825, l(2)k03514
Chromosome Location
Chr.2L: 8709880 - 8740660 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003209
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201804.1 NP_001188733.1 3 - 4 1124 75
NM_001298830.1 NP_001285759.1 2 - 3 574 75
NM_001298831.1 NP_001285760.1 2 - 3 1445 75
NM_080050.3 NP_524789.2 2 - 3 390 75
NM_164836.3 NP_723416.1 3 - 4 924 75
GenBank mRNA BT044207.1 - 2 - 3 361 75

View Full Product Details