Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
salm
Entrez Gene ID
34569
Gene Name
spalt major
Gene Aliases
3602, B1164, CG6464, Dmel\CG6464, Dmel_CG6464, SAL, SALM, SAL[M], SPALT, Sal, SalM, Salm, Spalt, l(2)03602, sal, salII, salM, slam, spalt
Chromosome Location
Chr.2L: 11434311 - 11445614 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261648
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298914.1 NP_001285843.1 2 - 3 4427 80
NM_164966.3 NP_723670.2 2 - 3 4427 80

View Full Product Details