Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncoa6
Entrez Gene ID
33761
Gene Name
Nuclear receptor coactivator 6
Gene Aliases
CG1402, CG14023, Dmel\CG14023, Dmel_CG14023, NCOA6, NcoA6, dNCOA6
Chromosome Location
Chr.2L: 5315290 - 5326683 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031698
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103618.3 NP_001097088.3 5 - 6 7456 62
NM_001258955.2 NP_001245884.1 5 - 6 7456 62
NM_135077.3 NP_608921.3 5 - 6 7453 62

View Full Product Details