Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Neurl4
- Entrez Gene ID
-
39558
- Gene Name
- Neuralized E3 ubiquitin protein ligase 4
- Gene Aliases
-
Blue, CG46284, CG6451, Dmel\CG46284, Dmel_CG46284, blue
- Chromosome Location
-
Chr.3L: 14043694 - 14049892 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0283503
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140431.3 |
NP_648688.2 |
6 - 7 |
3772 |
69 |
View Full Product Details