Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sytbeta
- Entrez Gene ID
-
39630
- Gene Name
- Synaptotagmin beta
- Gene Aliases
-
CG13462, CG3020, CG42333, Dmel\CG42333, Dmel_CG42333, SYT-beta, Syt V, Syt beta, Syt13, SytV, dsytbeta, syt13, sytbeta
- Chromosome Location
-
Chr.3L: 15014695 - 15064977 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261090
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140477.2 |
NP_648734.1 |
2 - 3 |
706 |
72 |
View Full Product Details