Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbs
Entrez Gene ID
49070
Gene Name
Myosin binding subunit
Gene Aliases
0573/06, 0953/04, CG32156, CG5600, CG5891, DMBS, DMYPT, DMbs, Dmel\CG32156, Dmel_CG32156, MBS, Mypt, l(3)03802, l(3)72Dd, l(3)S005331b, l(3)S057306, l(3)S057306b, l(3)S095304, l(3)j3B4, l(3)j7A1, mbs
Chromosome Location
Chr.3L: 16051959 - 16082026 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005536
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274990.1 NP_001261919.1 17 - 18 3708 69
NM_001382056.1 NP_001369054.1 5 - 6 1597 69
NM_140573.6 NP_648830.1 6 - 7 1132 69
GenBank mRNA AY051508.1 - 6 - 7 1098 69

View Full Product Details