Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Atg3
- Entrez Gene ID
-
40044
- Gene Name
- Autophagy-related 3
- Gene Aliases
-
ATG3, AUT1, Aut-1, Aut1, CG6877, Dmel\CG6877, Dmel_CG6877, Draut1, atg3, dAtg3
- Chromosome Location
-
Chr.3L: 18748687 - 18750483 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036813
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140802.3 |
NP_649059.1 |
1 - 2 |
207 |
73 |
GenBank mRNA |
AY119557.1 |
- |
1 - 2 |
186 |
73 |
View Full Product Details