Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Max
Entrez Gene ID
40095
Gene Name
CG9648-PA
Gene Aliases
CG9648, Dmel\CG9648, Dmel_CG9648, MAX, bHLHd9, dMax, dmax, max
Chromosome Location
Chr.3L: 19262232 - 19264184 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0017578
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259904.2 NP_001246833.1 4 - 5 505 77
NM_001300185.1 NP_001287114.1 4 - 5 505 77
NM_140840.4 NP_649097.1 4 - 5 505 77
GenBank mRNA BT025211.1 - 4 - 5 513 77
KX532135.1 - 4 - 5 346 77
U77369.1 - 4 - 5 395 77

View Full Product Details