Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tey
Entrez Gene ID
40146
Gene Name
teyrha-meyrha
Gene Aliases
CG8780, Dmel\CG8780, Dmel_CG8780
Chromosome Location
Chr.3L: 19650506 - 19664104 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036899
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144509.3 NP_001137981.1 2 - 3 659 72
NM_001275130.2 NP_001262059.1 2 - 3 629 72
NM_001275131.1 NP_001262060.1 2 - 3 474 72
NM_140883.2 NP_649140.1 2 - 3 659 72
GenBank mRNA BT133456.1 - 2 - 3 658 72

View Full Product Details