Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mTerf3
- Entrez Gene ID
-
40279
- Gene Name
- mitochondrial transcription termination factor 3
- Gene Aliases
-
CG5047, D-MTERF3, DmMTERF3, DmMterf3, Dmel\CG5047, Dmel_CG5047, MTERF3, Mterf3
- Chromosome Location
-
Chr.3L: 20519590 - 20520772 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037008
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140983.3 |
NP_649240.1 |
1 - 2 |
821 |
72 |
GenBank mRNA |
AY051744.1 |
- |
1 - 2 |
793 |
72 |
View Full Product Details