Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grp
Entrez Gene ID
34993
Gene Name
grapes
Gene Aliases
1C, CG17161, CHK1, CHK1/Grapes, Chk1, Dchk1, DmChk1, Dmel\CG17161, Dmel_CG17161, Grp, Grp/DChk1, Pk36A, Pk?1, chk1, dChk1, grp(chk), grps, l(2)k15102, lemp
Chromosome Location
Chr.2L: 16680054 - 16699926 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261278
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057663.4 NP_477011.1 3 - 4 1066 109
NM_165171.3 NP_723985.1 3 - 4 737 109
NM_165172.3 NP_723986.1 4 - 5 847 109
GenBank mRNA AY051961.1 - 3 - 4 1041 109

View Full Product Details