Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
blot
Entrez Gene ID
39939
Gene Name
bloated tubules
Gene Aliases
CG3897, DmBLOT, Dmel\CG3897, Dmel_CG3897, l(3)1658
Chromosome Location
Chr.3L: 17396031 - 17421514 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027660
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079401.3 NP_524125.2 2 - 3 1240 55
NM_170636.2 NP_730263.1 2 - 3 810 55
GenBank mRNA BT133440.1 - 2 - 3 1190 55

View Full Product Details