Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
blot
Entrez Gene ID
39939
Gene Name
bloated tubules
Gene Aliases
CG3897, DmBLOT, Dmel\CG3897, Dmel_CG3897, l(3)1658
Chromosome Location
Chr.3L: 17396031 - 17421514 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027660
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300164.1 NP_001287093.1 3 - 4 511 52
NM_079401.3 NP_524125.2 3 - 4 1476 52
NM_170636.2 NP_730263.1 3 - 4 1046 52
NM_170637.2 NP_730264.1 2 - 3 349 52
NM_170638.2 NP_730265.1 2 - 3 479 52
GenBank mRNA BT033062.1 - 2 - 3 349 52
BT133104.1 - 3 - 4 514 52
BT133440.1 - 3 - 4 1426 52

View Full Product Details