Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bchs
- Entrez Gene ID
-
33819
- Gene Name
- blue cheese
- Gene Aliases
-
Alfy, BcDNA:SD12984, Bchs, Beach1, CG14001, CG9011, DBEACH1, Dmel\CG14001, Dmel_CG14001, EP2299, alfy, bchd, beach
- Chromosome Location
-
Chr.2L: 5907169 - 5922776 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0043362
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135124.2 |
NP_608968.2 |
11 - 12 |
8805 |
98 |
View Full Product Details