Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pez
Entrez Gene ID
33882
Gene Name
CG9493-PA
Gene Aliases
CG9493, Dmel\CG9493, Dmel_CG9493, PEZ, PTPD1, dPez
Chromosome Location
Chr.2L: 6332881 - 6338671 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031799
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273198.1 NP_001260127.1 6 - 7 3688 65
NM_135169.3 NP_609013.1 5 - 6 3597 65
GenBank mRNA AF217284.1 - 5 - 6 3555 65

View Full Product Details