Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ent2
Entrez Gene ID
33921
Gene Name
Equilibrative nucleoside transporter 2
Gene Aliases
BEST:LD04971, CG11045, CG31911, DmENT2, DmEnt2, Dmel\CG31911, Dmel_CG31911, ENT2, LD04971, P124, ent2
Chromosome Location
Chr.2L: 6562325 - 6564709 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263916
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298751.1 NP_001285680.1 1 - 1 1776 139
NM_135205.4 NP_609049.1 1 - 1 1776 139
GenBank mRNA AY058347.1 - 1 - 1 1750 139

View Full Product Details