Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slob
Entrez Gene ID
34038
Gene Name
Slowpoke binding protein
Gene Aliases
CG13792, CG43756, CG6772, Dmel\CG43756, Dmel_CG43756, slob
Chromosome Location
Chr.2L: 7642005 - 7675078 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264087
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201784.2 NP_001188713.1 12 - 13 3544 86
NM_001273282.1 NP_001260211.1 12 - 13 3336 86
NM_135287.2 NP_609131.2 11 - 12 3011 86

View Full Product Details