Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdc14
Entrez Gene ID
34067
Gene Name
cell division cycle 14
Gene Aliases
CDC-14, CDC14, CG7134, Cdc14, Dmel\CG7134, Dmel_CG7134
Chromosome Location
Chr.2L: 7801668 - 7810703 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031952
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169433.2 NP_001162904.1 3 - 4 903 65
NM_001169434.2 NP_001162905.1 3 - 4 903 65
NM_001298792.1 NP_001285721.1 3 - 4 903 65
NM_001298793.1 NP_001285722.1 3 - 4 903 65
NM_135309.3 NP_609153.1 3 - 4 903 65
GenBank mRNA BT016136.1 - 3 - 4 614 65

View Full Product Details