Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hand
- Entrez Gene ID
-
34379
- Gene Name
- CG18144-PA
- Gene Aliases
-
CG18144, Dhand, Dmel\CG18144, Dmel_CG18144, HAND, bHLHa28, dHAND, dHand, hand, hand-C
- Chromosome Location
-
Chr.2L: 10292679 - 10295385 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032209
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135526.3 |
NP_609370.2 |
3 - 4 |
527 |
105 |
GenBank mRNA |
BT126289.1 |
- |
3 - 4 |
528 |
105 |
View Full Product Details