Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpr19
Entrez Gene ID
34408
Gene Name
defective proboscis extension response 19
Gene Aliases
BcDNA:LP09705, CG13140, Dmel\CG13140, Dmel_CG13140, Dpr-19, Dpr19
Chromosome Location
Chr.2L: 10376168 - 10379905 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032233
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273407.2 NP_001260336.1 2 - 3 530 152
NM_001273408.2 NP_001260337.1 3 - 4 339 152
NM_135548.3 NP_609392.1 2 - 3 551 152
GenBank mRNA AY122217.1 - 2 - 3 537 152

View Full Product Details