Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
swm
Entrez Gene ID
35235
Gene Name
second mitotic wave missing
Gene Aliases
CG10084, Dmel\CG10084, Dmel_CG10084, E1, EC2-10, S(Rux)2B, S(rux)2B, Swm, l(2)37Dh, l(2)E1
Chromosome Location
Chr.2L: 19493251 - 19497978 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002044
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273657.1 NP_001260586.1 2 - 3 420 94
NM_001273658.1 NP_001260587.1 1 - 2 386 94
NM_136132.5 NP_609976.1 2 - 3 420 94
NM_165304.2 NP_724201.1 1 - 2 386 94
GenBank mRNA AY095056.1 - 2 - 3 407 94
EF601876.1 - 2 - 3 409 94

View Full Product Details