Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sNPF
- Entrez Gene ID
-
35286
- Gene Name
- short neuropeptide F precursor
- Gene Aliases
-
38B.2, CG13968, Dmel\CG13968, Dmel_CG13968, Drm-sNPF, Drm-sNPF-1, Drm-sNPF-2, Drm-sNPF-3, Drm-sNPF-4, Drm-sNPF-AP1, LRLRFamide, SNPF, lrlrfa, s-NPF, sNPF-1, sNPF-1[4-11], sNPF-2, sNPF-2[12-19], sNPF-2s, sNPF-3, sNPF-4, sNPF1, sNPF1[1-11], sNPF1[4-11, sNPF2[12-19], sNPF3, short NPF, snpf
- Chromosome Location
-
Chr.2L: 20027915 - 20056373 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032840
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165316.2 |
NP_724239.1 |
3 - 4 |
1020 |
64 |
GenBank mRNA |
BT150152.1 |
- |
3 - 4 |
1020 |
64 |
View Full Product Details