Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lok
Entrez Gene ID
35288
Gene Name
loki
Gene Aliases
10895, 38B.4, CG10895, CHK-2, CHK2, Chk2, DmCHk2, DmChk2, Dmchk2, Dmel\CG10895, Dmel_CG10895, Dmnk, Dmnk-L, Dmnk-S, Dmnk/DChk2, LOKI/CHK2, Lok, Loki, MNK, Mnk, chk, chk-2, chk2, chk2/lok, mnk
Chromosome Location
Chr.2L: 20061422 - 20064381 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0019686
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057870.4 NP_477218.1 5 - 6 1161 68
NM_057871.4 NP_477219.1 5 - 6 1287 68
NM_165318.3 NP_724241.1 4 - 5 1222 68
GenBank mRNA AY070549.1 - 5 - 6 1116 68

View Full Product Details