Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr39a
- Entrez Gene ID
-
117346
- Gene Name
- Gustatory receptor 39a
- Gene Aliases
-
39D.2, 39D.2a, 39D.2b, 39D.2c, 39D.2d, 39D2a, 39D2b, 39D2c, 39D2d, CG31622, Dmel\CG31622, Dmel_CG31622, GR39D.2, GR39a, Gr39D2A, Gr39D2a, Gr39D2b, Gr39D2c, Gr39D2d, Gr39a.a, Gr39a.b, Gr39a.c, Gr39a.d, Gr39aA, Gr39aB, Gr39aC, Gr39aD, gene Y, l(2)k05106
- Chromosome Location
-
Chr.2L: 21290734 - 21297686 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264556
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165371.1 |
NP_724330.1 |
2 - 3 |
958 |
147 |
View Full Product Details