Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-6
Entrez Gene ID
35971
Gene Name
CG11804-PA
Gene Aliases
CED-6, CG11804, Ced-6, Dmel\CG11804, Dmel_CG11804, ced6, dCed-6, dced-6, dced6, drCed-6, drCed6, drced-6
Chromosome Location
Chr.2R: 9434669 - 9444937 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029092
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259296.2 NP_001246225.1 3 - 4 1016 60
NM_001259297.2 NP_001246226.2 3 - 4 1126 60
NM_136644.3 NP_610488.1 3 - 4 811 60
NM_165675.3 NP_724789.1 3 - 4 936 60
NM_165676.2 NP_724790.1 3 - 4 1126 60
GenBank mRNA AF204777.1 - 3 - 4 845 60
AY119596.1 - 3 - 4 844 60
BT125651.1 - 3 - 4 1017 60

View Full Product Details