Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gem
Entrez Gene ID
36064
Gene Name
gemini
Gene Aliases
BcDNA:HL07889, CG11867, CG30011, CG3459, Dmel\CG30011, Dmel_CG30011, anon-AE003830.1, dCP2
Chromosome Location
Chr.2R: 10124293 - 10137472 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050011
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136712.4 NP_610556.1 5 - 6 4102 62
NM_165748.3 NP_724893.3 5 - 6 2444 62
NM_165750.4 NP_724895.1 5 - 6 2860 62
GenBank mRNA AY075312.1 - 5 - 6 3010 62

Gene Symbol
CG46319
Entrez Gene ID
30522408
Gene Name
CG46319-PA
Gene Aliases
Dmel\CG46319, Dmel_CG46319, gem-5'utr
Chromosome Location
Chr.2R: 10124293 - 10137472 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347798.1 NP_001334715.1 5 - 6 2860 62
NM_001347799.1 NP_001334716.1 5 - 6 4102 62
NM_001347800.1 NP_001334717.1 5 - 6 2444 62

View Full Product Details