Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nub
Entrez Gene ID
34669
Gene Name
nubbin
Gene Aliases
CG15488, CG15489, CG34395, CG6246, Dmel\CG34395, Dmel_CG34395, NUB, Nb, Nub, Oct-1, Oct1, PDM-1, PDM1, POU33F1, Pdm, Pdm-1, Pdm1, dOct1, dPOU-19, dPOU-19/pdm-1, dPOU19, nb, pdm, pdm-1, pdm1, twn
Chromosome Location
Chr.2L: 12587625 - 12628135 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085424
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103683.2 NP_001097153.1 2 - 2 815 63

View Full Product Details