Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mipp2
- Entrez Gene ID
-
31544
- Gene Name
- Multiple inositol polyphosphate phosphatase 2
- Gene Aliases
-
CG4317, Dmel\CG4317, Dmel_CG4317, l(1)G0050, l(1)G0303, mipp2
- Chromosome Location
-
Chr.X: 6082709 - 6091471 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026060
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078500.4 |
NP_511055.1 |
2 - 3 |
784 |
67 |
GenBank mRNA |
AY095178.1 |
- |
2 - 3 |
701 |
67 |
View Full Product Details